Morpholin-3-one 98% (by GC)

Supplier: Ambeed
Warning

A153909-100G A153909-10G A153909-500G A153909-25G A153909-5G
103842-760EA 8 USD
103842-760 76719-836 76719-838 76719-840 77318-446 77318-448
Morpholin-3-one 98% (by GC)
Morpholin-3-one

Precaución: for Research Use Only

Formula: C₄H₇NO₂
MW: 101.1039 g/mol
Boiling Pt: 313 °C (760 mm Hg)
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00631009
CAS Number: 109-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR