Benzyltrimethylammonium tribromide 97%

Supplier: Ambeed
Warning

Synonyms: Mono(N,N,N-trimethyl-1-phenylmethanaminium) tribromide, Benzyl trimethyl ammonium tribromide

A137746-100G A137746-10G A137746-1G A137746-500G A137746-25G A137746-5G
77109-634EA 36.08 USD
77109-634 77109-636 77109-638 77109-640 77315-480 77315-482
Benzyltrimethylammonium tribromide 97%
Benzyltrimethylammonium tribromide

Precaución: for Research Use Only

Formula: C₁₀H₁₆Br₃N
MW: 389.97 g/mol
Melting Pt: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00134423
CAS Number: 111865-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR