Mono-6-O-(p-toluenesulfonyl)-β-cyclodextrin 95%

Supplier: Ambeed
Warning

Synonyms: Mono-(6-p-toluenesulphonyl)-β-cyclodextrin

A447247-1G A447247-100MG A447247-250MG A447247-5G
103827-758EA 86 USD
103827-758 76842-624 77156-102 77156-104 77156-106
Mono-6-O-(p-toluenesulfonyl)-β-cyclodextrin 95%
Mono-6-O-(p-toluenesulfonyl)-beta-cyclodextrin

Precaución: for Research Use Only

Formula: C₄₉H₇₆O₃₇S
Storage Temperature: Refrigerator
MDL Number: MFCD05864975
CAS Number: 67217-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR