Moc-Val-OH 98%

Supplier: Ambeed
Warning

A112069-100G A112069-10G A112069-25G A112069-500G A112069-5G
103808-086EA 173 USD
103808-086 103823-780 76809-968 76809-970 76809-972 76809-974 77284-208
Moc-Val-OH 98%
Moc-Val-OH

Precaución: for Research Use Only

Formula: C₇H₁₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD12795357
CAS Number: 74761-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR