3-Methoxyphenol 95%

Supplier: Ambeed
Warning

A538395-25G A538395-100G A538395-10G A538395-500G A538395-5G
103839-732EA 5 USD
103839-732 76843-772 77103-418 77103-420 77103-422 77307-762
3-Methoxyphenol 95%
3-Methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₈O₂
Boiling Pt: 244 °C (760 mm Hg)
Melting Pt: …18 °C
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00002267
CAS Number: 150-19-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR