Mitoxantrone 98%

Supplier: Ambeed
Danger

Synonyms: 1,4-Dihydroxy-5,8-bis[[2-[(2-hydroxyethyl)amino]ethyl]amino]anthracene-9,10-dione, 1,4-Dihydroxy-5,8-bis((2-((2-hydroxyethyl)amino)ethyl)amino)anthracene-9,10-dione, 1,4-Dihydroxy-5,8-bis[[2-[(2-hydroxyethyl)amino]ethyl]amino]anthraquinone

A207951-100MG A207951-1G A207951-1MG A207951-250MG A207951-5G
103792-406EA 19 USD
103792-406 103799-486 76721-630 76721-632 76721-634 76721-636 76721-638
Mitoxantrone 98%
Mitoxantrone

Precaución: for Research Use Only

Formula: C₂₂H₂₈N₄O₆
MW: 444.49 g/mol
Boiling Pt: 805.7 °C (760 mm Hg)
Melting Pt: 170…174 °C
Flash Pt: 441.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00242942
CAS Number: 65271-80-9
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR