Methylboronic acid 98%

Supplier: Ambeed
Warning

A110324-10G A110324-25G A110324-1G A110324-5G A110324-100G A110324-1000G A110324-500G
103797-652EA 5 USD
103797-652 103823-460 77270-842 77270-844 77283-974 77283-976 77549-134 77614-846 77614-848
Methylboronic acid 98%
Methylboronic acid

Precaución: for Research Use Only

Formula: CH₅BO₂
MW: 59.86 g/mol
Melting Pt: 91…94 °C
Storage Temperature: Freezer
MDL Number: MFCD00002105
CAS Number: 13061-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR