Methyl-2-bromo-2-(2-chlorophenyl)acetate 98%

Supplier: Ambeed
Danger

A831883-100G A831883-500G A831883-25G
103841-168EA 10 USD
103841-168 76728-752 76728-754 76845-404
Methyl-2-bromo-2-(2-chlorophenyl)acetate 98%
Methyl-2-bromo-2-(2-chlorophenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₈BrClO₂
Boiling Pt: 281 °C (760 mm Hg)
Flash Pt: 124 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08444353
CAS Number: 85259-19-4
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR