Methyl trans-4-aminocyclohexanecarboxylate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: trans-4-Aminocyclohexanecarboxylic Acid Methyl Ester Hydrochloride

A109857-100G A109857-10G A109857-250MG A109857-25G A109857-1G A109857-5G
103797-580EA 15 USD
103797-580 103823-384 76970-664 76970-666 76970-668 76970-670 77283-902 77310-188
Methyl trans-4-aminocyclohexanecarboxylate hydrochloride 97%
Methyl trans-4-aminocyclohexanecarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₅NO₂
MW: 193.67 g/mol
Melting Pt: 250…254 °C
Flash Pt: 108.3 °C
Storage Temperature: Ambient
MDL Number: MFCD08274538
CAS Number: 61367-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR