Methyl thiophene-3-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Thiophene-3-carboxylic Acid Methyl Ester

A142735-100G A142735-10G A142735-1G A142735-25G A142735-5G
103803-206EA 5 USD
103803-206 76972-678 76972-680 77316-450 77316-452 77316-454
Methyl thiophene-3-carboxylate 98%
Methyl thiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆O₂S
MW: 142.17 g/mol
Boiling Pt: 192.5 °C (760 mm Hg)
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD06203810
CAS Number: 22913-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR