Methyl quinoxaline-5-carboxylate 98%

Supplier: Ambeed
Warning

A155337-250MG A155337-1G A155337-5G
103804-746EA 47 USD
103804-746 103842-942 76812-428 77318-658 77318-660
Methyl quinoxaline-5-carboxylate 98%
Methyl quinoxaline-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD19703859
CAS Number: 6924-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR