Quinoxaline-2-carboxylic acid methyl ester 98%
Supplier: Ambeed
|
Warning
|
A263647-10G
A263647-250MG
A263647-1G
A263647-5G
A263647-25G
103812-634EA
98
USD
103812-634
103852-524
76827-168
76827-170
77092-168
77092-170
77304-876
Quinoxaline-2-carboxylic acid methyl ester 98%
Quinoxaline-2-carboxylic acid methyl ester
Formula:
C₁₀H₈N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD02167640 CAS Number: 1865-11-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...