Methyl quinoline-2-carboxylate 98%

Supplier: Ambeed
Warning

A150963-10G A150963-250MG A150963-1G A150963-25G A150963-5G
103804-222EA 34 USD
103804-222 103842-366 76719-714 76719-716 77317-900 77317-902 77317-904
Methyl quinoline-2-carboxylate 98%
Methyl quinoline-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉NO₂
Boiling Pt: 321.2±15.0 °C (760 mm Hg)
Flash Pt: 148 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00160630
CAS Number: 19575-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR