Methyl 3-pyrrolidinecarboxylate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl pyrrolidine-3-carboxylate hydrochloride
A125824-100G
A125824-10G
A125824-1G
A125824-5G
A125824-25G
103800-728EA
16
USD
103800-728
77094-740
77094-742
77298-088
77298-090
77312-876
Methyl 3-pyrrolidinecarboxylate hydrochloride 97%
Methyl 3-pyrrolidinecarboxylate hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₁NO₂·ClH MW: 165.618 g/mol Boiling Pt: 213.6 °C (760 mm Hg) Flash Pt: 83 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08436169 CAS Number: 198959-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...