Methyl pyrimidine-2-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A117964-100G
A117964-10G
A117964-25G
A117964-1G
A117964-5G
103798-964EA
6
USD
103798-964
103824-854
77108-368
77108-370
77108-372
77311-638
77311-640
Methyl pyrimidine-2-carboxylate 97%
Methyl pyrimidine-2-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₆N₂O₂ Boiling Pt: 251.3 °C (760 mm Hg) Flash Pt: 105.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD03426929 CAS Number: 34253-03-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...