Methyl propiolate 98%

Supplier: Ambeed
Danger

Synonyms: Methyl acetylenecarboxylate, Propiolic acid methyl ester, 2-Propynoic acid methyl ester

A380933-1G A380933-10G
103826-684EA 14 USD
103826-684 103837-718 77306-920 77551-110
Methyl propiolate 98%
Methyl propiolate

Precaución: for Research Use Only

Formula: C₄H₄O₂
Boiling Pt: 104 °C (760 mm Hg)
Flash Pt: 16 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008572
CAS Number: 922-67-8
UN: 3272
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR