Methyl-4-piperidinecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-Piperidinecarboxylate, Methyl isonipecotate, Methyl piperidine-4-carboxylate

A128242-10G A128242-5G A128242-25G A128242-100G A128242-500G A128242-1000G
77109-028EA 9.55 USD
77109-028 77109-030 77313-392 77549-528 77611-120 77685-110
Methyl-4-piperidinecarboxylate 97%
Methyl 4-piperidinecarboxylate

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MW: 143.18 g/mol
Boiling Pt: 193.8 °C (760 mm Hg)
Flash Pt: 89 °C
Storage Temperature: Freezer
MDL Number: MFCD00190578
CAS Number: 2971-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR