Methyl-3-piperidinecarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl piperidine-3-carboxylate

A234386-25G A234386-5G A234386-10G
103850-702EA 5 USD
103850-702 77089-688 77089-690 77113-106
Methyl-3-piperidinecarboxylate 95%
Methyl 3-piperidinecarboxylate

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
Boiling Pt: 193.8 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD00797850
CAS Number: 50585-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR