Methyl pipecolinate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Methyl Pipecolinate HCl, H-DL-Pip-OMe·HCl, Methyl piperidine-2-carboxylate hydrochloride, Methyl 2-piperidinecarboxylate hydrochloride, Pipecolinic acid methyl ester hydrochloride, 2-Piperidinecarboxylic acid methyl ester hydrochloride

A169875-25G A169875-5G A169875-100G
103844-688EA 6 USD
103844-688 77081-784 77301-008 77667-372
Methyl pipecolinate hydrochloride 97%
Methyl pipecolinate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₄ClNO₂
Boiling Pt: 235.3 °C (760 mm Hg)
Flash Pt: 96.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00192316
CAS Number: 32559-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR