Methyl picolinate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl pyridine-2-carboxylate
A230718-5G
A230718-10G
A230718-100G
A230718-500G
A230718-1G
103850-468EA
7
USD
103850-468
77089-340
77112-984
77615-918
77615-920
77685-632
Methyl picolinate 97%
Methyl picolinate
Precaución: for Research Use Only
Formula:
C₇H₇NO₂ MW: 137.14 g/mol Boiling Pt: 227.4 °C (760 mm Hg) Flash Pt: 103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038038 CAS Number: 2459-07-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...