Methyl oxetane-3-carboxylate 95%

Supplier: Ambeed
Warning

A169820-1G A169820-100MG A169820-250MG
103806-418EA 139 USD
103806-418 103844-682 77081-778 77151-228 77151-230
Methyl oxetane-3-carboxylate 95%
Methyl oxetane-3-carboxylate

Precaución: for Research Use Only

Formula: C₅H₈O₃
Boiling Pt: 126.0±15.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD22566365
CAS Number: 1638760-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR