Methyl-N-methylanthranilate 98%
Supplier: Ambeed
|
Warning
|
A498263-100G
A498263-10G
A498263-500G
A498263-25G
A498263-1000G
103839-356EA
7
USD
103839-356
76804-944
76804-946
76804-948
76843-326
77626-988
Methyl-N-methylanthranilate 98%
Methyl-N-methylanthranilate
Precaución: for Research Use Only
Formula:
C₉H₁₁NO₂ Boiling Pt: 252.4±13.0 °C (760 mm Hg) Melting Pt: 17…19 °C Flash Pt: 106.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017183 CAS Number: 85-91-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...