Methyl-N-cbz-4-piperidineacetate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl N-Cbz-4-piperidineacetate

A243857-250MG A243857-1G A243857-25G
103811-552EA 29 USD
103811-552 103851-376 76976-544 77304-288 77304-290
Methyl-N-cbz-4-piperidineacetate 95%
Methyl-N-cbz-4-piperidineacetate

Precaución: for Research Use Only

Formula: C₁₆H₂₁NO₄
MW: 291.3400 g/mol
Boiling Pt: 402.9±28.0 °C (760 mm Hg)
Flash Pt: 197.5 °C
Storage Temperature: Ambient
MDL Number: MFCD02179006
CAS Number: 170737-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR