Methyl-N-boc-piperidine-3-carboxylate 97%

Supplier: Ambeed
Warning

A154626-100G A154626-500G A154626-25G A154626-5G
103808-804EA 434 USD
103808-804 103842-860 77010-946 77010-948 77318-596 77318-598
Methyl-N-boc-piperidine-3-carboxylate 97%
Methyl-N-boc-piperidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
Boiling Pt: 307.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06795926
CAS Number: 148763-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR