Methyl-N-boc-4-piperidinepropionate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl N-Boc-4-piperidinepropionate

A116262-250MG A116262-1G
103798-700EA 26 USD
103798-700 103824-562 76718-314 77311-380
Methyl-N-boc-4-piperidinepropionate 97%
Methyl-N-boc-4-piperidinepropionate

Precaución: for Research Use Only

Formula: C₁₄H₂₅NO₄
MW: 271.3526 g/mol
Boiling Pt: 339.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD02179013
CAS Number: 162504-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR