Methyl morpholine-2-carboxylate hydrochloride 97%

Supplier: Ambeed
Warning

A299143-10G A299143-250MG A299143-1G
103814-216EA 16 USD
103814-216 103835-362 76816-724 76816-726 76838-058
Methyl morpholine-2-carboxylate hydrochloride 97%
Methyl morpholine-2-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD17167182
CAS Number: 937063-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR