Methyl isoquinoline-3-carboxylate 98%

Supplier: Ambeed
Warning

A284741-10G A284741-250MG A284741-1G A284741-5G
103813-650EA 44 USD
103813-650 103834-782 76802-274 76802-276 76837-308 76837-310
Methyl isoquinoline-3-carboxylate 98%
Methyl isoquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉NO₂
MW: 187.2000 g/mol
Boiling Pt: 344.6 °C (760 mm Hg)
Melting Pt: 84…88 °C
Flash Pt: 162.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00075138
CAS Number: 27104-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR