Methyl isoindoline-4-carboxylate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A147530-100MG
A147530-250MG
A147530-50MG
A147530-1G
A147530-5G
103803-792EA
89
USD
103803-792
76871-488
76871-490
76871-492
77317-272
77883-166
Methyl isoindoline-4-carboxylate hydrochloride 97%
Methyl isoindoline-4-carboxylate hydrochloride
Precaución: for Research Use Only
Formula:
C₁₀H₁₂ClNO₂ MW: 213.67 g/mol Boiling Pt: 345.6 °C (760 mm Hg) Flash Pt: 162.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD10700133 CAS Number: 127168-90-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...