Methyl indoline-5-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A147635-100MG
A147635-250MG
A147635-1G
A147635-5G
103803-810EA
59
USD
103803-810
103841-918
76719-600
76719-602
77317-310
77317-312
Methyl indoline-5-carboxylate 98%
Methyl indoline-5-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₁NO₂ MW: 177.1998 g/mol Boiling Pt: 330.9 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD07371630 CAS Number: 141452-01-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...