Methyl indole-4-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A107365-100G
A107365-10G
A107365-25G
A107365-1G
A107365-5G
103797-180EA
6
USD
103797-180
103822-948
77270-652
77270-654
77270-656
77309-796
77309-798
Methyl indole-4-carboxylate 98%
Methyl indole-4-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₉NO₂ MW: 175.19 g/mol Boiling Pt: 331.7 °C (760 mm Hg) Melting Pt: 68…71 °C Flash Pt: 154.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191222 CAS Number: 39830-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...