Methyl imidazo[1,2-a]pyridine-5-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A100679-250MG
A100679-1G
103795-568EA
55
USD
103795-568
103821-264
76809-206
77296-798
Methyl imidazo[1,2-a]pyridine-5-carboxylate 98%
Methyl imidazo[1,2-a]pyridine-5-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₈N₂O₂ MW: 176.1700 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12026020 CAS Number: 88047-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...