Methyl-1-cyclohexene-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl cyclohex-1-enecarboxylate

A187837-100G A187837-10G A187837-1G A187837-25G A187837-5G
103816-046EA 13 USD
103816-046 103846-640 76799-914 76799-916 77302-102 77302-104 77302-106
Methyl-1-cyclohexene-1-carboxylate 98%
Methyl-1-cyclohexene-1-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₂O₂
MW: 140.182 g/mol
Boiling Pt: 190…192 °C (760 mm Hg)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00001544
CAS Number: 18448-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR