cis-4-Hydroxy-L-proline methyl ester hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-cis-Hyp-OMe·HCl, Methyl cis-4-Hydroxy-L-proline Hydrochloride

A382474-100MG A382474-250MG A382474-1G A382474-5G A382474-100G A382474-10G
103826-714EA 16 USD
103826-714 103837-750 76803-732 76803-734 77306-934 77306-936 77668-780 77879-825
cis-4-Hydroxy-L-proline methyl ester hydrochloride 97%
cis-4-Hydroxy-L-proline methyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00237835
CAS Number: 40126-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR