Methyl bromopyruvate 80%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-Bromopyruvate, Methyl 3-Bromo-2-oxopropionate, Bromopyruvic acid methyl ester, 3-Bromopyruvic acid methyll ester, 3-Bromo-2-oxopropionic acid methyll ester, 3-Bromo-2-oxopropionic acid methyl ester

A145016-100G A145016-1G A145016-25G A145016-5G
103841-560EA 7 USD
103841-560 76871-412 76871-414 77316-832 77316-834
Methyl bromopyruvate 80%
Methyl bromopyruvate

Precaución: for Research Use Only

Formula: BrH₂CC(O)COOCH₃
Flash Pt: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD01862963
CAS Number: 7425-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR