Methyl-1,3-benzoxazole-5-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A182419-100MG
A182419-250MG
A182419-1G
103807-710EA
38
USD
103807-710
103846-082
76998-728
76998-730
77301-782
Methyl-1,3-benzoxazole-5-carboxylate 97%
Methyl-1,3-benzoxazole-5-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₇NO₃ Melting Pt: 110…111 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08689687 CAS Number: 924869-17-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...