Methyl benzenesulfinate 97%

Supplier: Ambeed
Warning

A273602-100G A273602-10G A273602-1G A273602-25G A273602-5G
103813-148EA 9 USD
103813-148 103853-070 76723-102 76723-104 77092-866 77092-868 77092-870
Methyl benzenesulfinate 97%
Methyl benzenesulphinate

Precaución: for Research Use Only

Formula: C₇H₈O₂S
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00674051
CAS Number: 670-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR