Methyl azetidine-3-carboxylate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Azetidine-3-carboxylic acid methyll ester hydrochloride, Methyl Azetidine-3-carboxylate Hydrochloride, Azetidine-3-carboxylic methyl ester hydrochloride

A354265-1G A354265-100G A354265-10G A354265-25G A354265-5G
103826-118EA 6 USD
103826-118 103837-122 76840-242 77033-132 77033-134 77306-658 77306-660
Methyl azetidine-3-carboxylate hydrochloride 97%
Methyl azetidine-3-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀ClNO₂
MW: 151.59 g/mol
Boiling Pt: 190.9 °C (760 mm Hg)
Flash Pt: 69.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01861758
CAS Number: 100202-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR