Methyl-2-azetidinecarboxylate hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Methyl azetidine-2-carboxylate hydrochloride

A244686-100MG A244686-250MG A244686-1G
103811-594EA 62 USD
103811-594 103851-428 76801-396 76801-398 77304-328
Methyl-2-azetidinecarboxylate hydrochloride 95%
Methyl-2-azetidinecarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀ClNO₂
MW: 151.59 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01861757
CAS Number: 162698-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR