Methyl-ɑ-D-mannopyranoside 95%

Supplier: Ambeed
Warning

Synonyms: ɑ-Methyl-D-mannoside, Methyl-ɑ-D-Mannoside, Methyl α-D-Mannopyranoside, ɑ-Methyl-D-mannopyranoside, (2R,3S,4S,5S,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol

A132708-10G A132708-25G A132708-100G A132708-500G A132708-1000G
76719-012EA 6 USD
76719-012 77314-358 77566-580 77566-582 77878-953
Methyl-ɑ-D-mannopyranoside 95%
Methyl-alpha-D-mannopyranoside

Precaución: For research use only.

Formula: C₇H₁₄O₆
MW: 194.18 g/mol
Boiling Pt: 418.236 °C (760 mm Hg)
Flash Pt: 206.742 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063262
CAS Number: 617-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR