Methyl 7-nitrobenzo[b]thiophene-2-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A101815-100MG
A101815-1G
103796-152EA
94
USD
103796-152
76982-192
77282-838
Methyl 7-nitrobenzo[b]thiophene-2-carboxylate 95%
Methyl 7-nitrobenzo[b]thiophene-2-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₇NO₄S Boiling Pt: 393.2 °C (760 mm Hg) Flash Pt: 191.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD13180959 CAS Number: 34084-89-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...