Methyl-7-bromoheptanoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 7-bromoheptanoate

A127988-100G A127988-10G A127988-1G A127988-25G A127988-5G
103801-050EA 20 USD
103801-050 76718-832 76718-834 77313-336 77313-338 77313-340
Methyl-7-bromoheptanoate 98%
Methyl-7-bromoheptanoate

Precaución: for Research Use Only

Formula: C₈H₁₅BrO₂
Boiling Pt: 229.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD02258672
CAS Number: 54049-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR