Methyl 7-aminoheptanoate hydrochloride 97%

Supplier: Ambeed
Warning

A108643-10G A108643-5G A108643-1G
103797-376EA 5 USD
103797-376 103823-156 76970-570 77283-762 77309-996
Methyl 7-aminoheptanoate hydrochloride 97%
Methyl 7-aminoheptanoate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₇NO₂·ClH
MW: 195.687 g/mol
Melting Pt: 119…121 °C
Flash Pt: 105.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09029389
CAS Number: 17994-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR