Adipic semialdehyde methyl ester 97%

Supplier: Ambeed
Warning

Synonyms: 6-Oxohexanoic Acid Methyl Ester, Methyl 6-oxohexanoate, Hexanoic acid,6-oxo-,methyl ester

A179488-250MG A179488-1G A179488-5G A179488-10G A179488-25G
103807-434EA 42 USD
103807-434 103845-782 77011-934 77301-614 77301-616 77683-440 77683-441
Adipic semialdehyde methyl ester 97%
Adipic semialdehyde methyl ester

Precaución: For research use only.

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 189.4 °C (760 mm Hg)
Flash Pt: 71 °C
Storage Temperature: Freezer
MDL Number: MFCD00063422
CAS Number: 6654-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR