Methyl 6-oxoheptanoate 98%

Supplier: Ambeed
Warning

A243618-250MG A243618-1G A243618-25G
103811-526EA 29 USD
103811-526 103851-348 76826-796 77090-550 77090-552
Methyl 6-oxoheptanoate 98%
Methyl 6-oxoheptanoate

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MW: 158.2 g/mol
Boiling Pt: 227.3 °C (760 mm Hg)
Flash Pt: 91.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22376711
CAS Number: 2046-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR