Methyl 6-oxo-3,6-dihydropyrimidine-4-carboxylate 98%

Supplier: Ambeed
Warning

A225755-250MG A225755-1G
103819-196EA 30 USD
103819-196 103850-096 76815-034 77088-802
Methyl 6-oxo-3,6-dihydropyrimidine-4-carboxylate 98%
Methyl 6-oxo-3,6-dihydropyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
MW: 154.12 g/mol
Boiling Pt: 263.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD21640297
CAS Number: 7399-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR