Methyl 6-oxo-1,6-dihydropyridazine-4-carboxylate 95%

Supplier: Ambeed
Warning

A195279-100MG A195279-250MG
103816-734EA 53 USD
103816-734 103847-348 76691-054 76691-056
Methyl 6-oxo-1,6-dihydropyridazine-4-carboxylate 95%
Methyl 6-oxo-1,6-dihydropyridazine-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD19707403
CAS Number: 89640-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR