Methyl-3-hydroxypyridazine-6-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-oxo-1,6-dihydropyridazine-3-carboxylate, Methyl 1,6-dihydro-6-oxopyridazine-3-carboxylate

A777086-1G A777086-25G A777086-5G A777086-100G A777086-10G A777086-250MG
103840-954EA 19 USD
103840-954 76845-212 76845-214 76845-216 77006-380 77006-382 77006-384
Methyl-3-hydroxypyridazine-6-carboxylate 97%
Methyl-3-hydroxypyridazine-6-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17015975
CAS Number: 63001-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR