Methyl-6-methylpicolinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-methylpyridine-2-carboxylate

A110733-100G A110733-10G A110733-25G A110733-1G A110733-5G
103797-714EA 9 USD
103797-714 103823-522 76781-056 76781-058 76781-060 77310-324 77310-326
Methyl-6-methylpicolinate 98%
Methyl 6-methylpicolinate

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.17 g/mol
Boiling Pt: 242 °C (760 mm Hg)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD06203670
CAS Number: 13602-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR