Methyl 6-methylpyrazolo[1,5-a]pyridine-3-carboxylate 97%

Supplier: Ambeed
Warning

A102103-100MG A102103-250MG
103796-300EA 662 USD
103796-300 77148-250 77148-252
Methyl 6-methylpyrazolo[1,5-a]pyridine-3-carboxylate 97%
Methyl 6-methylpyrazolo[1,5-a]pyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12405113
CAS Number: 127717-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR