Methyl-6-methylnicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-methylnicotinate

A192590-100G A192590-10G A192590-500G A192590-5G A192590-25G A192590-1000G
103809-276EA 147 USD
103809-276 76690-936 76690-938 76690-940 76690-942 77084-944 77302-306
Methyl-6-methylnicotinate 98%
Methyl-6-methylnicotinate

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 228.4 °C (760 mm Hg)
Melting Pt: 34…37 °C
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00006340
CAS Number: 5470-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR